Learn More
HRH3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533846
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is not recommended for use in WB applications. Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Mouse, Rat, Hamster, Elephant, Bovine, Horse, Guinea pig (100%).
HRH3 Polyclonal antibody specifically detects HRH3 in Equine, Guinea Pig, Hamster, Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
HRH3 | |
Polyclonal | |
Unconjugated | |
HRH3 | |
AW049250; G protein-coupled receptor; G protein-coupled receptor 97; GPCR97; G-protein coupled receptor 97; H3R; HH3R; histamine H3 receptor; histamine H3 receptor H3L isoform; histamine H3 receptor H3S isoform; histamine receptor; histamine receptor H 3; histamine receptor H3; Hrh3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100058016, 100135541, 100764853, 11255, 85268, 99296 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P58406, Q9JI35, Q9QYN8, Q9Y5N1 | |
HRH3 | |
Synthetic 16 amino acid peptide from 2nd cytoplasmic domain of human HRH3 / Histamine H3 Receptor. | |
50 μg | |
Primary | |
Equine, Guinea Pig, Hamster, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |