missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2A.X Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA523450
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Histone H2A.X Polyclonal antibody specifically detects Histone H2A.X in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Histone H2A.X | |
Polyclonal | |
Unconjugated | |
H2AX | |
AW228881; gamma H2AX; gammaH2ax; H2A histone family member X; H2A histone family, member X; H2A.X; H2A.X variant histone; H2A/X; h2afx; H2ax; H2AX histone; Hist5-2ax; histone 5 protein 2ax; histone H2A.x; Histone H2a/x; Histone H2AX; RGD1566119; similar to H2A histone family, member X; zgc:56329 | |
Rabbit | |
Protein A | |
RUO | |
15270, 3014 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
P16104, P27661 | |
H2AX | |
A portion of amino acids 50-110 of human Histone H2A.x | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |