Learn More
Histone H2B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA141058
Weitere Details : Netto-Gewicht : 1.24740kg
Beschreibung
This peptide sequence is 100% conserved between multiple species including human, mouse, rat, dog, chimpanzee, cow, chicken, duck, zebrafish, and drosophila. Suggested positive control: Jurkat, PBMC.
Histone H2B Polyclonal antibody specifically detects Histone H2B in Canine, Chicken, Drosophila, Human, Mammalia, Mouse, Rat, Zebrafish samples. It is validated for ChIP Assay, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Histone H2B | |
Polyclonal | |
Unconjugated | |
LOC102724334 | |
2610022J01Rik; Acetyl-Histone H2; AV127319; bA317E16.3; CG17949; CG17949-PA; CG33868; CG33868-PA; CG33870; CG33870-PA; CG33872; CG33872-PA; CG33874; CG33874-PA; CG33876; CG33876-PA; CG33878; CG33878-PA; CG33880; CG33880-PA; CG33882; CG33882-PA; CG33884; CG33884-PA; CG33886; CG33886-PA; CG33888; CG33888-PA; CG33890; CG33890-PA; CG33892; CG33892-PA; CG33894; CG33894-PA; CG33896; CG33896-PA; CG33898; CG33898-PA; CG33900; CG33900-PA; CG33902; CG33902-PA; CG33904; CG33904-PA; CG33906; CG33906-PA; CG33908; CG33908-PA; CG33910; CG33910-PA; dJ160A22.3; dJ193B12.2; dJ221C16.3; dJ221C16.6; dJ221C16.8; dJ97D16.4; Dmel\CG17949; Dmel\CG33868; Dmel\CG33870; Dmel\CG33872; Dmel\CG33874; Dmel\CG33876; Dmel\CG33878; Dmel\CG33880; Dmel\CG33882; Dmel\CG33884; Dmel\CG33886; Dmel\CG33888; Dmel\CG33890; Dmel\CG33892; Dmel\CG33894; Dmel\CG33896; Dmel\CG33898; Dmel\CG33900; Dmel\CG33902; Dmel\CG33904; Dmel\CG33906; Dmel\CG33908; Dmel\CG33910; Dmel_CG17949; Dmel_CG33868; Dmel_CG33870; Dmel_CG33872; Dmel_CG33874 | |
Rabbit | |
Protein G | |
RUO | |
100329290, 100329560, 100333496, 100334559, 100334869, 100858607, 102724334, 114483833, 128312, 158983, 24829, 255626, 286436, 291157, 3017, 3018, 319177, 319178, 319181, 319182, 319183, 319184, 319186, 319190, 326273, 361247, 3771809, 3771891, 3771957, 3772013, 3772058, 3772081, 3772083, 3772094, 3772099, 3772104, 3772166, 3772203, 3772248, 3772264, 3772265, 3772271, 3772276, 3772299, 3772336, 3772496, 3772502, 3772575, 386920, 426037, 440689, 478743, 482202, 488252, 491396, 608682, 68024, 680312, 691488, 8339, 8340, 8341, 8342, 8345, 8346, 8347, 8348, 8349, 85236, 8970 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ChIP Assay, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
A0A2R8Y619, O60814, P06899, P0C1H6, P10854, P23527, P33778, P57053, P58876, P62807, P70696, Q00729, Q16778, Q5QNW6, Q64475, Q64478, Q64524, Q6PC60, Q6ZWY9, Q7Z2G1, Q8CGP1, Q8N257, Q93079, Q96A08, Q99877, Q99879, Q99880 | |
H2B, H2bc1, H2BC10, H2BC11, H2BC12, H2BC13, H2BC14, H2BC15, H2BC17, H2bc18, H2BC21, H2BC3, H2BC4, H2BC5, H2BC8, H2BC9, H2BE1, H2BU1, H2BW1, H2BW2, His2B:CG17949, His2B:CG33868, His2B:CG33870, His2B:CG33872, His2B:CG33874, His2B:CG33876, His2B:CG33878, His2B:CG33880, His2B:CG33882, His2B:CG33884, His2B:CG33886, His2B:CG33888, His2B:CG33890, His2B:CG33892, His2B:CG33894, His2B:CG33896, His2B:CG33898, His2B:CG33900, His2B:CG33902, His2B:CG33904, His2B:CG33906, His2B:CG33908, His2B:CG33910, HIST1H2BA, HIST1H2BD, HIST1H2BJ, HIST1H2BM, HIST1H2BO, hist2h2l, LOC100329290, LOC100329560, LOC100334559, LOC100334869, LOC102724334, LOC426037, LOC478743 | |
Synthetic peptide corresponding to amino acids 111-125 of human histone H2B [GenBank no. CAI19747.1] | |
100 μg | |
Primary | |
Canine, Chicken, Drosophila, Human, Mammalia, Mouse, Rat, Zebrafish | |
Antibody | |
IgG |