Learn More
Histone H3.3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527507
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: IMR32, PC-12, Rat2. Predicted reactivity: Mouse (100%), Zebrafish (99%), Xenopus laevis (100%), Pig (100%), Rabbit (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Histone H3.3 Polyclonal antibody specifically detects Histone H3.3 in Human, Mouse, Rat samples. It is validated for ChIP Assay, Immunocytochemistry, Western Blot.Spezifikation
Histone H3.3 | |
Polyclonal | |
Unconjugated | |
H3-3A | |
9430068D06Rik; CG5825; CG5825-PA; CG5825-PC; CG5825-PD; CG8989; dH3.3A; Dmel\CG5825; Dmel_CG5825; family 3B (H3.3B); H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3.3; H3.3 histone A; H3.3 histone B; H3.3A; H3.3A/B; H3.3B; H3.4; H3.A; H3.B; H3/g; H3-3A; H3-3B; H3B; H3F3; H3F3A; H3F3A protein; H3F3B; H3FT; H3-IX; H3L-like histone; H3t; H3-X; His3.3; His3.3A; His-3.3A; His3.3A-PA; His3.3A-PC; His3.3A-PD; His3.3B; histone; Histone cluster 3; Histone H3; histone H3 class II; Histone H3.3; histone H3.3A; Histone H3.3B; histone H3.3-like protein; MGC126886; MGC126888; PH3; PP781; similar to H3 histone, family 3A; unnamed protein product | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3020, 3021 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ChIP Assay, Immunocytochemistry, Western Blot | |
0.08 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P84243 | |
H3-3A, H3-3B | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 105 of Human Histone H3.3 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |