missing translation for 'onlineSavingsMsg'
Learn More

H4ac pan-acetyl (K5,K8,K12,K16) Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA184526

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11559170

  • 600.00 € / 100 Mikroliter
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Rabbit anti Histone H4 (Acetylated) antibody was raised against the tetra-acetylated form of histone H4 and recognizes acetylated isoforms of histone H4.

H4ac pan-acetyl (K5,K8,K12,K16) Polyclonal antibody specifically detects H4ac pan-acetyl (K5,K8,K12,K16) in All, Human, Mouse, Rat samples. It is validated for ChIP Assay, ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot.
Spezifikation

Spezifikation

H4ac pan-acetyl (K5,K8,K12,K16)
Polyclonal
Unconjugated
H4C1
acetyl Histone H4; Acetyl-H4 K5 K8 K12 K16; Acetyl-H4 Lys5 Lys8 Lys12 Lys16; Acetyl-Histone H4 K5 K8 K12 K16; Acetyl-Histone H4 Lys5 Lys8 Lys12 Lys16; FO108; H4; H4 ac pan-acetyl; H4 histone family, member A; H4 histone family, member I; H4 histone family, member J; H4 histone family, member M; H4 histone family, member N; H4 histone, family 2; H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/J; H4/K; H4/M; H4/n; H4/O; H4/p; H4-12; H4-16; H4-53; H4ac; H4c1; H4c11; H4C12; H4C13; H4c14; H4C15; H4c2; H4c3; H4c4; H4C5; H4c6; H4C8; H4c9; H4f16; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; H4K12; H4K12a; H4K12ac; H4K16; H4K16ac; H4K5; H4K5,8,12,16ac; H4K5ac; H4K8; H4K8ac; H4Lys5,8,12,16ac; H4M; HIST1H4; HIST1H4A; Hist1h4b; HIST1H4C; Hist1h4d; HIST1H4E; Hist1h4f; Hist1h4h; Hist1h4i; HIST1H4J; Hist1h4k; HIST1H4L; Hist1h4m; Hist2h4; HIST2H4A; HIST2H4B; Hist4h4; histone 1, H4a; histone 1, H4b; histone 1, H4e; histone 1, H4i; histone 2, H4; histone 2, H4a;
Rabbit
100 μL
Primary
All species, Human, Mouse, Rat
Antibody
IgG
ChIP Assay, ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Conc. Not Determined
whole serum with <0.1% sodium azide
P62805, P62806
H4-16, H4C1, H4C11, H4C14, H4C2, H4C5, H4C9
KLH-conjugated peptide acetylated at lysines 5,8,12,16: NSGRGAcKGGAcKGLGAcKGGAAcKRHYCC
RUO
121504, 319158, 326619, 326620, 8294, 8359, 8363, 8366, 8367, 8370, 97122
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote