Learn More
HIV1 gp120 Polyclonal Antibody, Biotin, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA173098
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The PA1-73098 antibody reacts with HIV-1 gp120. PA1-73098 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-73098 antibody was raised against Purified native gp120 from strain IIIB.
HIV1 gp120 Polyclonal antibody specifically detects HIV1 gp120 in Virus samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Spezifikation
HIV1 gp120 | |
Polyclonal | |
Biotin | |
HIV Glycoprotein 120; HIV-1 Glycoprotein 120; Human immunodeficiency virus | |
Purified native gp120 from strain IIIB. | |
1 mL | |
Primary | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
4-5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Goat | |
purified | |
RUO | |
Virus | |
Antibody | |
IgG |