missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIV1 nef Monoclonal Antibody (2H12), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA171505
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody reacts with HIV-1 Nef, B and C subtypes, BRU, IIIB, and HAN strains. This clone may also be referred to as 2H12 (01-007).
HIV1 nef Monoclonal antibody specifically detects HIV1 nef in Virus samples. It is validated for ELISA, Western Blot.Spezifikation
HIV1 nef | |
Monoclonal | |
0.6 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
2H12 | |
Unconjugated | |
HIV-1 Nef | |
Recombinant Nef protein of HIV-1 BRU purified from E. coli | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |