Learn More
HIV1 p24 Polyclonal Antibody, Biotin, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA173095
Weitere Details : Netto-Gewicht : 1.24740kg
Beschreibung
The PA1-73095 antibody reacts with HIV-1 p24. PA1-73095 has been successfully used in Western blot applications. The PA1-73095 antibody was raised against purified native p24 from strain IIIB. Recommended short term storage of less than six months, keep at 2-8oC. For long term storage, store at -20oC.
HIV1 p24 Polyclonal antibody specifically detects HIV1 p24 in Virus samples. It is validated for Western Blot.Spezifikation
HIV1 p24 | |
Polyclonal | |
Biotin | |
HIV p24; HIV1gp1 | |
Purified native p24 from strain IIIB. | |
1 mL | |
Primary | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
4-5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Goat | |
purified | |
RUO | |
Virus | |
Antibody | |
IgG |