Learn More
HIV1 p24 Monoclonal Antibody (G82B), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA17040
Weitere Details : Netto-Gewicht : 1.24740kg
Beschreibung
MA1-7040 detects the HIV-1 p24. MA1-7040 has been successfully used in sandwich ELISA procedures as a capture antibody, with PA1-73095 as the detection antibody. Also used in immunofluorescence applications. The MA1-7040 immunogen is purified recombinant HIV-1 p24.
HIV1 p24 Monoclonal antibody specifically detects HIV1 p24 in Virus samples. It is validated for ELISA, Immunocytochemistry.Spezifikation
HIV1 p24 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry | |
G82B | |
Unconjugated | |
HIV p24; HIV1gp1 | |
Purified recombinant HIV-1 p24. | |
1 mL | |
Primary | |
4° C | |
Liquid |