missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIV1 p24 Monoclonal Antibody (N17), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA171516
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody reacts with HIV-1 p24 subtypes B, C and A, and has been mapped to the C-terminus, aa 165- 231. This clone may also be referred to as N17 (05-004).
HIV1 p24 Monoclonal antibody specifically detects HIV1 p24 in Virus samples. It is validated for ELISA, Western Blot.Spezifikation
HIV1 p24 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
N17 | |
Unconjugated | |
HIV p24; HIV1gp1 | |
Recombinant p24 protein of HIV-1 B-subtype consensus purified from E. coli | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |