Learn More
HIV1 p24 Polyclonal Antibody, HRP, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA173094
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The PA1-73094 antibody reacts with HIV-1 p24. PA1-73094 has been successfully used in ELISA applications. The PA1-73094 antibody was raised against purified native p24 from strain IIIB. NOTE: sodium azide is a potent inhibitor of peroxidase.
HIV1 p24 Polyclonal antibody specifically detects HIV1 p24 in Virus samples. It is validated for ELISA.Spezifikation
HIV1 p24 | |
Polyclonal | |
HRP | |
HIV p24; HIV1gp1 | |
Purified native p24 from strain IIIB. | |
1 mL | |
Primary | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |
ELISA | |
1-2 mg/mL | |
PBS with 10mg/mL BSA and 0.002% thimerosal | |
Goat | |
purified | |
RUO | |
Virus | |
Antibody | |
IgG |