Learn More
HIV1 Tat Monoclonal Antibody (7), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA171509
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody reacts with HIV-1 Tat BH10, IIIB strains. This antibody reacts with the rTat consensus sequences of HIV-1 subtypes A (PNLEPWH), B (PRLEPWK) and C (PNLEPWN). It has been mapped to amino acids 6-12 (PRLEPWK - B subtype a.a. consensus). MA1-71509 can be used for chromatin immunoprecipitation (ChIP) analysis of HIV1 Tat. This clone may also be referred to as 7 (02-002).
HIV1 Tat Monoclonal antibody specifically detects HIV1 Tat in Virus samples. It is validated for ChIP Assay, ELISA, Western Blot.Spezifikation
HIV1 Tat | |
Monoclonal | |
1.0 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ChIP Assay, ELISA, Western Blot | |
7 | |
Unconjugated | |
HIV-1 Tat | |
Recombinant Tat protein of HIV-1 BH10 purified from E. coli | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |