missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIV1 Vif Monoclonal Antibody (319), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA518030
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This target has a predicted molecular weight of 23 kDa.
HIV1 Vif Monoclonal antibody specifically detects HIV1 Vif in Virus samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
HIV1 Vif | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Mouse | |
Protein G | |
RUO | |
Virus | |
Antibody | |
IgG1 κ |
Immunocytochemistry, Western Blot | |
319 | |
Unconjugated | |
HIV-1 | |
Bacterially expressed, hexahistidine amino-terminal tagged HIV-1 Vif protein (clade B, HXB-3 isolate) | |
100 μL | |
Primary | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |