Learn More
HIV1 Virions Polyclonal Antibody, Biotin, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA173092
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The PA1-73092 antibody reacts with HIV-1 Virions. Does not react wtih T4+ human lymphocytes. PA1-73092 has been successfully used in Immunofluorescence, immunohistochemistry, ELISA, and western blot applications. The PA1-73092 antibody was raised against HIV-1 isolate. Store at 4oC short term. For long term storage, store at -20oC, avoiding freeze/thaw cycles.
HIV1 Virions Polyclonal antibody specifically detects HIV1 Virions in Virus samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Spezifikation
HIV1 Virions | |
Polyclonal | |
Biotin | |
HIV-1; HIV-1 Virions | |
Human confirmed HIV-1 isolate. | |
1 mL | |
Primary | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
4-5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Goat | |
purified | |
RUO | |
Virus | |
Antibody | |
IgG |