missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serum Albumin Monoclonal Antibody (15C7), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA190420
Weitere Details : Netto-Gewicht : 1.24740kg
Beschreibung
MA1-90420 detects HSA from human samples.
Human Serum Albumin Monoclonal antibody specifically detects Human Serum Albumin in Human samples. It is validated for ELISA, Western Blot.Spezifikation
Human Serum Albumin | |
Monoclonal | |
5.6 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P02768 | |
ALB | |
Full length protein (Human) | |
1 mg | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA, Western Blot | |
15C7 | |
Unconjugated | |
ALB | |
Alb; Alb1; Alb-1; albumin; albumin (32 AA); albumin (AA 34); albumin 1; Albza; ANALBA; cell growth inhibiting protein 42; DKFZp779N1935; FDAH; GIG20; GIG42; growth-inhibiting protein 20; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341 | |
Mouse | |
Protein A | |
RUO | |
213 | |
4° C | |
Liquid |