missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA526800
Detalles adicionales : Netto-Gewicht : 0.01000kg
Descripción
This antibody is predicted to react with non-human primate based on sequence homology.
HSD17B11 Polyclonal antibody specifically detects HSD17B11 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Especificaciones
HSD17B11 | |
Polyclonal | |
Unconjugated | |
HSD17B11 | |
17-beta-HSD 11; 17-beta-HSD XI; 17betaHSD11; 17-BETA-HSD11; 17beta-HSD11; 17betaHSDXI; 17-BETA-HSDXI; 17-beta-hydroxysteroid dehydrogenase 11; 17-beta-hydroxysteroid dehydrogenase type XI; 17-beta-hydroxysteroid dehydrogenase XI; 17bHSD11; CTCL tumor antigen HD-CL-03; CTCL-associated antigen HD-CL-03; Cutaneous T-cell lymphoma-associated antigen HD-CL-03; dehydrogenase/reductase (SDR family) member 8; dehydrogenase/reductase SDR family member 8; Dhrs8; estradiol 17-beta-dehydrogenase 11; estradiol 17-beta-dehydrogenase 11-like protein; Hsd17b11; hydroxysteroid (17-beta) dehydrogenase 11; hydroxysteroid 17-beta dehydrogenase 11; MGC137132 protein; PAN1B; PSEC0029; Retinal short-chain dehydrogenase/reductase 2; retinal short-chain dehydrogenase/reductase SDR2; retSDR2; SDR16C2; SDR2; short chain dehydrogenase/reductase family 16C member 2; short chain dehydrogenase/reductase family 16C, member 2; UNQ207/PRO233 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
51170 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q8NBQ5 | |
HSD17B11 | |
KLH conjugated synthetic peptide between 81-110 amino acids from the N-terminal region of human HSD17B11 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |