missing translation for 'onlineSavingsMsg'
Learn More

HSD17B6 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA542018

Weitere Details : Netto-Gewicht : 0.00010kg

Artikelnummer. 15535192

  • 471.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Peptide sequence: MWLYLAAFVG LYYLLHWYRE RQVVSHLQDK YVFITGCDSG FGNLLARQLD Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 100%; Rabbit: 79%; Rat: 100%

HSD17B6 Polyclonal antibody specifically detects HSD17B6 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

HSD17B6
Polyclonal
Unconjugated
HSD17B6
17-beta-HSD 6; 17-beta-HSD6; 17betaHSD9; 17-beta-HSD9; 17-beta-hydroxysteroid dehydrogenase type 6; 3(alpha->beta)-hydroxysteroid epimerase; 3(alpha->beta)-hydroxysteroid epimerasel; 3-alpha->beta-HSE; 3-alpha->beta-hydroxysteroid epimerase; 3-hydroxysteroid epimerase; Gm182; Hsd17b6; Hsd17b9; HSE; hydroxysteroid (17-beta) dehydrogenase 6; hydroxysteroid (17-beta) dehydrogenase 6 homolog; hydroxysteroid (17-beta) dehydrogenase 9; hydroxysteroid 17-beta dehydrogenase 6; hydroxysteroid 17-beta dehydrogenase 9; hypothetical protein LOC533086; NAD+ -dependent 3 alpha-hydroxysteroid dehydrogenase 3-hydroxysteroid epimerase; oxidative 17 beta hydroxysteroid dehydrogenase type 6; oxidative 3-alpha hydroxysteroid dehydrogenase; oxidative 3-alpha-hydroxysteroid-dehydrogenase; oxidoreductase; Rdh8; retinol dehydrogenase; RODH; SDR9C6; Short chain dehydrogenase/reductase family 9C member 6; short chain dehydrogenase/reductase family 9C, member 6
Rabbit
Protein A
RUO
8630
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
O14756
HSD17B6
Synthetic peptide directed towards the N-terminal of human HSD17B6
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote