Learn More
HSF2BP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527939
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: NT2D1, SK-N-SH. Predicted reactivity: Mouse (92%), Rat (91%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HSF2BP Polyclonal antibody specifically detects HSF2BP in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HSF2BP | |
Polyclonal | |
Unconjugated | |
HSF2BP | |
4932437G14Rik; heat shock factor 2-binding protein; heat shock transcription factor 2 binding protein; Hsf2bp; RGD1565879 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11077, 74377 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.93 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O75031, Q9D4G2 | |
HSF2BP | |
Recombinant fragment corresponding to a region within amino acids 1 and 259 of Human HSF2BP | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |