missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP20 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538279
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
HSPB6 is associated with actin (see MIM 102540) and modulates smooth muscle relaxation.Spezifikation
HSP20 | |
Polyclonal | |
Unconjugated | |
HSPB6 | |
Hsp20, heat shock 20 kDa-like protein p20, heat shock protein beta-6, HSPB6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126393, 192245, 243912 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O14558, Q5EBG6, P97541 | |
HSPB6 | |
A synthetic peptide derived from the internal region of human HSP20 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |