Learn More
HSP701L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529212
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse testis, rat testis, U87-MG, SK-N-SH, IMR32, SK-N-AS. Predicted reactivity: Mouse (95%), Rat (96%), Xenopus laevis (81%), Pig (96%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HSP701L Polyclonal antibody specifically detects HSP701L in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HSP701L | |
Polyclonal | |
Unconjugated | |
Hspa1l | |
heat shock 10kDa protein 1-like; heat shock 70 kDa protein 1-Hom; heat shock 70 kDa protein 1L; Heat shock 70 kDa protein 1-like; Heat shock 70 kDa protein 3; heat shock 70 kDa-like protein 1; heat shock 70kD protein-like 1; heat shock 70kDa protein 1-like; Heat shock protein; heat shock protein 1-like; Heat shock protein 70-1l; heat shock protein cognate 70, testis; heat shock protein family A (Hsp70) member 1 like; heat shock-related protein hsc70t; Hsc70t; HSP70.3; HSP70-1L; Hsp70-3; HSP70-HOM; HSP70T; Hspa1l; hum70t; inducible heat shock protein 70; Msh5; Spermatid-specific heat shock protein 70 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15482, 24963, 3305 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P16627, P34931, P55063 | |
Hspa1l | |
Recombinant fragment corresponding to a region within amino acids 11 and 278 of Human HSP70 1L | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |