missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPA12A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA526768
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
HSPA12A Polyclonal antibody specifically detects HSPA12A in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HSPA12A | |
Polyclonal | |
PBS with 0.09% sodium azide | |
O43301, Q8K0U4 | |
HSPA12A | |
KLH conjugated synthetic peptide between 398-427 amino acids from the central region of human HSPA12A | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
HSPA12A | |
1700063D12Rik; AI118035; AI840429; AW048913; AW556406; D5Mgi40; heat shock 70 kDa protein 12A; heat shock 70kD protein 12A; heat shock 70kDa protein 12A; Heat shock protein; heat shock protein 12A; heat shock protein family A (Hsp70) member 12A; HSP; HSPA12A; KIAA0417; mKIAA0417 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
259217, 73442 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |