Learn More
HSPA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529083
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A549, H1299, HCT116, MCF-7, NIH-3T3, BCL-1, U87-MG, SK-N-SH, IMR32, SK-N-AS, PC-12, Rat-2, rat brain. Predicted reactivity: Mouse (99%), Rat (98%), Xenopus laevis (92%), Chicken (96%), Chimpanzee (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HSPA2 Polyclonal antibody specifically detects HSPA2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HSPA2 | |
Polyclonal | |
Unconjugated | |
HSPA2 | |
70 kDa heat shock protein; 70kDa; Hcp70.2; Heat shock 70 kDa protein 2; Heat shock 70 kDa protein 3; heat shock 70kD protein 2; heat shock 70kDa protein 2; Heat shock protein; heat shock protein 2; heat shock protein 70.2; heat shock protein alpha 2; heat shock protein family A (Hsp70) member 2; heat shock protein, 70 kDa 2; heat shock-related 70 kDa protein 2; HSP; HSP70.2; HSP70.3; Hsp70-2; HSP70-3; HSP70A2; Hspa2; Hspt70; HST; Hst70; LOW QUALITY PROTEIN: heat shock-related 70 kDa protein 2; Testis-specific heat shock protein-related; testis-specific heat shock protein-related gene hst70 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15512, 3306, 60460 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
P14659, P17156, P54652 | |
HSPA2 | |
Recombinant fragment corresponding to a region within amino acids 295 and 578 of Human HSPA2 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |