missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2T Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518122
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
UBE2T Polyclonal antibody specifically detects UBE2T in Human samples. It is validated for Western Blot.Spezifikation
UBE2T | |
Polyclonal | |
Unconjugated | |
UBE2T | |
2700084L22Rik; C80607; Cell proliferation-inducing gene 50 protein; E2 ubiquitin-conjugating enzyme T; FANCT; HSPC150; HSPC150 protein similar to ubiquitin-conjugating enzyme; PIG50; RGD1310816; Ube2t; Ubiquitin carrier protein T; ubiquitin conjugating enzyme E2 T; ubiquitin conjugating enzyme E2T; ubiquitin-conjugating enzyme E2 T; ubiquitin-conjugating enzyme E2T; ubiquitin-conjugating enzyme E2T (putative); ubiquitin-protein ligase T | |
Goat | |
Antigen affinity chromatography | |
RUO | |
29089 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9NPD8 | |
UBE2T | |
Synthetic peptide sequence (QLVGIEKKFHPDV) corresponding to the C-terminus amino acids of UBE2T | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |