missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTR7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533314
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human (100%) Gorilla (94%) Gibbon (88%) Monkey (81%).
HTR7 Polyclonal antibody specifically detects HTR7 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
HTR7 | |
Polyclonal | |
Unconjugated | |
HTR7 | |
5-HT receptor 7a; 5-HT receptor 7b; 5Ht7; 5-HT7; 5-HT-7; 5HT7 Receptor; 5-HT7 receptor; 5-HT-X; 5-hydroxytryptamine (serotonin) receptor 7; 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled); 5-hydroxytryptamine (serotonin) receptor 7, adenylate cyclase-coupled; 5-hydroxytryptamine receptor 7; GPRFO; high affinity serotonin receptor (5HT7); HT7; Htr7; OTTHUMP00000020077; OTTHUMP00000020078; OTTHUMP00000020079; Serotonin receptor 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3363 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P34969 | |
HTR7 | |
Synthetic 16 amino acid peptide from C-terminus of human 5HT7 Receptor. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |