missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTR1D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533296
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset (100%) Panda, Dog, Horse, Guinea pig (94%).
HTR1D Polyclonal antibody specifically detects HTR1D in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
HTR1D | |
Polyclonal | |
Unconjugated | |
HTR1D | |
5 - Hydroxytryptamine (serotonin) receptor 1D; 5HT1D; 5-HT1D; 5-HT-1D; 5HT1D Receptor; 5-HT1d receptor; 5-HT-1D-alpha; 5-HT1DR; 5-hydroxytryptamine (serotonin) receptor 1D; 5-hydroxytryptamine (serotonin) receptor 1D beta; 5-hydroxytryptamine (serotonin) receptor 1D, G protein-coupled; 5-hydroxytryptamine receptor 1D; AI853647; Gpcr14; G-protein coupled receptor 14; HT1d receptor; HT1DA; Htr1d; HTR1DA; Htr1db; HTRL; RDC4; RP23-32L5.3; serotonin 1D alpha receptor; Serotonin receptor 1D | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3352 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P28221 | |
HTR1D | |
Synthetic 17 amino acid peptide from 2nd extracellular domain of human 5HT1D Receptor. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |