Learn More
HTR1F Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532864
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Orangutan, Gibbon (100%); Gorilla, Monkey, Bovine, Elephant, Horse, Rabbit, Guinea pig, Platypus (94%); Dog, Bat (88%); Rat, Panda (81%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
HTR1F Polyclonal antibody specifically detects HTR1F in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
HTR1F | |
Polyclonal | |
Unconjugated | |
HTR1F | |
5-HT-1E-beta; 5ht1f; 5-HT1F; 5-HT-1F; 5HT1F Receptor; 5-HT1f receptor; 5HT6; 5-hydroxytryptamine (serotonin) receptor 1E beta; 5-hydroxytryptamine (serotonin) receptor 1F; 5-hydroxytryptamine (serotonin) receptor 1F, G protein-coupled; 5-hydroxytryptamine receptor 1F; Htr1eb; HTR1EL; HTR1F; MR77; Serotonin receptor 1F; serotonin receptor gene (similar to Mm Htr1eb) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3355 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P30939 | |
HTR1F | |
Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human 5HT1F Receptor. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |