missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTR2C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533307
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon, Monkey, Marmoset, Elephant (100%) Horse, Pig (94%).
HTR2C Polyclonal antibody specifically detects HTR2C in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
HTR2C | |
Polyclonal | |
Unconjugated | |
HTR2C | |
5ht1c; 5-HT1C; 5-HT-1C; 5HT-1C; 5-HT1c receptor; 5-HT2C; 5-HT-2C; 5HT2C Receptor; 5-HT2C receptor; 5-HT2cR; 5HTR2C; 5-HTR2C; 5-hydroxytryptamine (serotonin) receptor 2C; 5-hydroxytryptamine (serotonin) receptor 2C, G protein-coupled; 5-hydroxytryptamine receptor 1C; 5-hydroxytryptamine receptor 2C; HGNC:5295; HTR1C; Htr2c; RP11-810O3.1; RP23-15B7.1; serotonin 1c receptor; serotonin 5-HT-1C receptor; serotonin 5-HT-2C receptor; serotonin receptor 2C; SR1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3358 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P28335 | |
HTR2C | |
Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human 5HT2C Receptor. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |