Learn More
Huntingtin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA1003
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
Neoepitope antibodies distinguish smaller cleaved fragments or processed forms of proteins versus the intact full-length or precursor by using a designed peptide purification process to maximize immunoreactivity to a specific cleavage site. Human HTT caspase cleavage sites generate fragment-specific forms of the protein. Caspase-3/7 has been shown to generate cleavage sites at animo acids 513 and 552. Caspase-2 cleaves at amino acid 552 and caspase-6 at amino acid 586. Neo-specific antibody PA1-003 recognizes the 552 cleaved fragment without detecting the full-length form.
Huntingtin Polyclonal antibody specifically detects Huntingtin in Human, Mouse samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot.Spezifikation
Huntingtin | |
Polyclonal | |
Unconjugated | |
Htt | |
AI256365; C430023I11Rik; Hd; HD protein; HD protein homolog; Hdh; HTT; Huntingtin; huntingtin (Huntington disease); Huntingtin, myristoylated N-terminal fragment; Huntington disease gene homolog; huntington disease protein; huntington disease protein homolog; IT15; solute carrier family 6 (neurotransmitter transporter, serotonin), member 4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
15194, 3064 | |
-20°C | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.4 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P42858, P42859 | |
Htt | |
Synthetic peptide conjugated to KLH via cysteine corresponding to residues SDPAMDLND (544-552) of Human HTT. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |