Learn More
APP Pan Specific Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF1168
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13416784
  • 351.13 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
APP Pan Specific Polyclonal antibody AF1168 Specifically detects APP Pan Specific in Human samples. It is validated for Immunohistochemistry, Western Blotting
Spezifikation
Spezifikation
APP
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
NP_958817
Amyloid beta, amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP, beta Amyloid, Protease Nexin II
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human APP695+1 and human APP770 in direct ELISAs and Western blots. This antibody recognizes a common epitope present on APP695+1 and APP770 found within the segments Leu18 - Arg288 or Pro365 - Arg411 of APP770 (Accession P05067).
Human
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
APP
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human APP 695+1 frameshift mutant, Leu18-Arg336, Accession # NP_958817
Immunogen affinity purified
RUO
Primary
351
Reconstitute at 0.2 mg/mL in sterile PBS.