Learn More
CDX4 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF2807
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417804
  • 351.13 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 30-07-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
CDX4 Polyclonal antibody AF2807 Specifically detects CDX4 in Human samples. It is validated for Immunohistochemistry, Western Blotting
Spezifikation
Spezifikation
CDX4
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
O14627
caudal type homeo box transcription factor 4, caudal type homeobox 4, caudal type homeobox transcription factor 4, Caudal-type homeobox protein 4, homeobox protein CDX-4
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human CDX4 in direct ELISAs and Western blots.
Human
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CDX4
E. coli-derived recombinant human CDX4 Met1-Thr170 Accession # O14627
Immunogen affinity purified
RUO
Primary
1046
Reconstitute at 0.2 mg/mL in sterile PBS.