Learn More
DCIR/CLEC4A Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF1748
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417134
  • 479.72 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
DCIR/CLEC4A Polyclonal antibody AF1748 Specifically detects DCIR/CLEC4A in Human samples. It is validated for Flow Cytometry, Western Blotting
Spezifikation
Spezifikation
DCIR/CLEC4A
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Q9UMR7
CD367, CLEC4A, CLECSF6, CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor, LLIR
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human DCIR/CLEC4A in direct ELISAs and Western blots.
Human
Western Blot, Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CLEC4A
Mouse myeloma cell line NS0-derived recombinant human DCIR/CLEC4A isoform 1, Gln70-Leu237, Accession # Q9UMR7
Immunogen affinity purified
RUO
Primary
50856
Reconstitute at 0.2 mg/mL in sterile PBS.