Learn More
Dishevelled-1 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF3316
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418104
  • 341.89 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Dishevelled-1 Polyclonal antibody AF3316 Specifically detects Dishevelled-1 in Human samples. It is validated for Immunohistochemistry, Western Blotting
Spezifikation
Spezifikation
Dishevelled-1
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
O14640
dishevelled 1 (homologous to Drosophila dsh), dishevelled, dsh homolog 1 (Drosophila), Dishevelled1, Dishevelled-1, Dsh, DSH homolog 1, DVL, DVL1, DVL1L1, MGC54245, segment polarity protein dishevelled homolog DVL-1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human Dishevelled-1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) DVL-2 and rhDVL-3 is observed.
Human
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
DVL1
E. coli-derived recombinant human Dishevelled-1, Arg150-Leu215, Accession # O14640
Immunogen affinity purified
RUO
Primary
1855
Reconstitute at 0.2 mg/mL in sterile PBS.