Learn More
EDA2R/TNFRSF27/XEDAR Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF1093
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13416734
  • 351.13 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 31-07-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
EDA2R/TNFRSF27/XEDAR Polyclonal antibody AF1093 Specifically detects EDA2R/TNFRSF27/XEDAR in Human samples. It is validated for Blocking (Receptor-ligand), Western Blotting
Spezifikation
Spezifikation
EDA2R/TNFRSF27/XEDAR
LYOPH
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction
Q9HAV5
ectodysplasin A2 receptor, EDA-A2 receptor, EDAA2R, EDA-A2R, EDAA2Rtumor necrosis factor receptor superfamily member XEDAR, EDAR2, TNFRSF27, XEDAR, XEDARTNFRSF27tumor necrosis factor receptor superfamily member 27, X-linked ectodysplasin-A2 receptor
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human EDA2R/TNFRSF27/XEDAR in direct ELISAs and Western blots.
Human
Western Blot, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
EDA2R
Mouse myeloma cell line NS0-derived recombinant human EDA2R/TNFRSF27/XEDAR Met1-Glu136 Accession # Q9HAV5
Immunogen affinity purified
RUO
Primary
60401
Reconstitute at 0.2 mg/mL in sterile PBS.