Learn More
FGF-6 Goat anti-Human, Polyclonal, R&D Systems™
Marke:  RD Systems (Bio-Techne) AF238
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417524
  • 351.13 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
FGF-6 Polyclonal antibody AF238 Specifically detects FGF-6 in Human samples. It is validated for Neutralization, Western Blotting
Spezifikation
Spezifikation
FGF-6
LYOPH
Western Blot 0.1 ug/mL, Neutralization 0.2-1 ug/mL
P10767
FGF6, FGF-6, fibroblast growth factor 6, HBGF-6, Heparin secretory-transforming protein 2, Heparin-binding growth factor 6, HST2, HST-2, HSTF2, HSTF-2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human FGF-6 in direct ELISAs and Western blots. In Western blots, approximately 50% cross-reactivity with recombinant human (rh) FGF-4 is observed, less than 5% cross-reactivity with rhFGF-9 is observed, and less than 2% cross-reactivity with FGF-acidic, rhFGF-5, and rhFGF-7 is observed. In direct ELISAs, less than 1% cross-reactivity with rh beta-ECGF, recombinant mouse (rm) FGF-8b, and rmFGF-8c is observed.
Human
Western Blot, Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FGF6
E. coli-derived recombinant human FGF-6, Gly41-Ile208, Accession # P10767
Immunogen affinity purified
RUO
Primary
2251
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos