Learn More
Frizzled-5 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF1617
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417054
  • 360.37 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 30-07-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Frizzled-5 Polyclonal antibody AF1617 Specifically detects Frizzled-5 in Human samples. It is validated for Western Blotting
Spezifikation
Spezifikation
Frizzled-5
LYOPH
Western Blot 0.1 ug/mL
AAC50385
C2orf31, chromosome 2 open reading frame 31, DKFZP434E2135, frizzled (Drosophila) homolog 5, frizzled homolog 5 (Drosophila), frizzled-5, Frizzled5, Frz-5, FZ-5, FZD5, FzE5, HFZ5, MGC129692, seven-transmembrane receptor frizzled-5, Wnt receptor
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Frizzled-5 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse (rm) Frizzled-1, rmFrizzled-2, rmFrizzled-3, rmFrizzled-6, rmFrizzled-7, and rmFrizzled-8 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FZD5
Mouse myeloma cell line NS0-derived recombinant human Frizzled-5 Ala27-Pro167 Accession # AAC50385
Immunogen affinity purified
RUO
Primary
7855
Reconstitute at 0.2 mg/mL in sterile PBS.