Learn More
Glypican 5 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF2607
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417674
  • 332.65 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 25-04-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Glypican 5 Polyclonal antibody AF2607 Specifically detects Glypican 5 in Human samples. It is validated for Western Blotting
Spezifikation
Spezifikation
Glypican 5
LYOPH
Western Blot 0.1 ug/mL
P78333
bA93M14.1, glypican 5, glypican proteoglycan 5, glypican-5, GPC5
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Glypican 5 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse Glypican 5 is observed and less than 2% cross-reactivity with recombinant human (rh) Glypican 2, rhGlypican 3, and rhGlypican 6 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
GPC5
Mouse myeloma cell line NS0-derived recombinant human Glypican 5 Glu25-Thr554 Accession # P78333
Immunogen affinity purified
RUO
Primary
2262
Reconstitute at 0.2 mg/mL in sterile PBS.