Learn More
gp130 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF-228-NA
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417464
  • 500.52 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 31-07-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
gp130 Polyclonal antibody AF228SP Specifically detects gp130 in Human samples. It is validated for Neutralization, Western Blotting
Spezifikation
Spezifikation
gp130/CD130
LYOPH
Western Blot 0.1 ug/mL, Neutralization 2-10 ug/mL
P40189
CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, IL6ST, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human gp130 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant rat gp130, recombinant mouse gp130, recombinant human (rh) IL-1 sRI, rhIL-1 sRII, rhIL-2 sR beta , rhIL-2 sR gamma , rhIL-4 sR, and rhIL-6 sR is observed.
Human
Western Blot, Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL6ST
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human gp130 Leu24-Glu619 (Glu619Asp) Accession # P40189
Immunogen affinity purified
RUO
Primary
3572
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos