Learn More
Growth Hormone Monoclonal Antibody (3E2), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA514682
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-14682 targets Human Growth Hormone in ELISA, IP, and RIA applications and shows reactivity with Human samples. The MA5-14682 immunogen is human Growth Hormone. MA5-14682 detects Human Growth Hormone which has a predicted molecular weight of approximately 24 kDa. Product MA514682 is a smaller package size of MIH9901 (formerly sold as a Seradyn product).
Growth Hormone Monoclonal antibody specifically detects Growth Hormone in Human samples. It is validated for ELISA, Immunoprecipitation, Radioimmune Assays (RIA).Spezifikation
Growth Hormone | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
P01241 | |
Gh1 | |
Human Growth Hormone | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
3E2 | |
Unconjugated | |
Gh1 | |
dog growth hormone; GH; Gh1; GHB1; GHB5; GHN; GH-N; Growth hormone; growth hormone 1; growth hormone 12; growth hormone B1; growth hormone B5; growth hormone precursor; HGH; hGH-N; IGHD1B; pituitary growth hormone; RNGHGP; Somatotropin | |
Mouse | |
Protein A | |
RUO | |
2688 | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |