missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Growth Hormone Monoclonal Antibody (GhB9), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA121650
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-21650 can be used in an ELISA pair with MA1-21649 (capture).
Growth Hormone Monoclonal antibody specifically detects Growth Hormone in Human samples. It is validated for ELISA.Spezifikation
Growth Hormone | |
Monoclonal | |
5.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P01241 | |
Gh1 | |
Recombinant full length protein (Human). | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
GhB9 | |
Unconjugated | |
Gh1 | |
dog growth hormone; GH; Gh1; GHB1; GHB5; GHN; GH-N; Growth hormone; growth hormone 1; growth hormone 12; growth hormone B1; growth hormone B5; growth hormone precursor; HGH; hGH-N; IGHD1B; pituitary growth hormone; RNGHGP; Somatotropin | |
Mouse | |
Protein G | |
RUO | |
2688 | |
4° C, do not freeze | |
Liquid |