Learn More
HAI-2 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF1106
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13416744
  • 351.13 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 22-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
HAI-2 Polyclonal antibody AF1106 Specifically detects HAI-2 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Spezifikation
Spezifikation
HAI-2/SPINT2
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
O43291
HAI2, HAI2A, HAI2B, HAI-2DIAR3, HAI2FLJ45571, Hepatocyte growth factor activator inhibitor type 2, KOP, kunitz-type protease inhibitor 2, PB, Placental bikunin, serine peptidase inhibitor, Kunitz type, 2, serine protease inhibitor, Kunitz type, 2, SPINT2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human HAI-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human HAI-1 and recombinant mouse HAI-2B is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SPINT2
Mouse myeloma cell line NS0-derived recombinant human HAI-2A, Ala28-Lys197, Accession # O43291
Immunogen affinity purified
RUO
Primary
10653
Reconstitute at 0.2 mg/mL in sterile PBS.