Learn More
IBSP/Sialoprotein II Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF4014
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418504
  • 360.37 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
IBSP/Sialoprotein II Polyclonal antibody AF4014 Specifically detects IBSP/Sialoprotein II in Human samples. It is validated for Western Blotting
Spezifikation
Spezifikation
IBSP/Sialoprotein II
LYOPH
Western Blot 0.1 ug/mL
AAC95490
BNSP, Bone sialoprotein, Bone sialoprotein 2, BSP, BSP 2, BSP II, BSP2, BSPII, BSP-II, Cell binding sialoprotein, IBSP, Integrin binding sialoprotein, SP II, SPII, SP-II
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human IBSP/Sialoprotein II in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IBSP
Chinese hamster ovary cell line CHO-derived recombinant human IBSP/Sialoprotein II, Phe17-Gln317, Accession # AAC95490
Immunogen affinity purified
RUO
Primary
3381
Reconstitute at 0.2 mg/mL in sterile PBS.