missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgA Mouse anti-Human, Invitrogen™
Mouse Monoclonal Secondary Antibody
Marke: Invitrogen LFMA0165
Weitere Details : Netto-Gewicht : 0.99000kg
Beschreibung
A suggested positive control for this product is human plasma.
Thermo Scientific Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Spezifikation
Human IgA | |
Monoclonal | |
1 mg/mL | |
Mouse | |
Protein G | |
RUO | |
Human | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
47C12 | |
Unconjugated | |
Protein purified from Human plasma | |
100 μL | |
Secondary | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 κ |