Learn More
Mouse anti-Human IgA (Heavy chain) Secondary Antibody, Invitrogen™
Mouse Monoclonal Secondary Antibody
Marke: Invitrogen MA180150
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Mouse anti Human IgA heavy chain antibody, clone Mc24-2E11 recognizes human immunoglobulin alpha heavy chains. No cross reactivity observed with IgG, IgM or free kappa and lambda light chains.
Human IgA (Heavy chain) Monoclonal antibody specifically detects Human IgA (Heavy chain) in Human samples. It is validated for ELISA, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Spezifikation
Human IgA (Heavy chain) | |
Monoclonal | |
1 mg/mL | |
TBS with 0.09% sodium azide; pH 7.4 | |
Reduced and alkylated human IgA. | |
200 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 |
ELISA, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
Mc24-2E11 | |
Unconjugated | |
Mouse | |
Protein A | |
RUO | |
Human | |
Liquid |