missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F(ab')2-Goat anti-Human IgA Secondary Antibody, Invitrogen™
F(ab')2-Goat Polyclonal Secondary Antibody
Marke: Invitrogen A24460
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Human IgA Polyclonal antibody specifically detects Human IgA in Human samples. It is validated for ELISA, Western Blot.Spezifikation
Human IgA | |
Polyclonal | |
Unconjugated | |
Goat | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
ELISA, Western Blot | |
> 4.5 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
Human IgA | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |