missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgE Secondary Antibody, AP, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen A18796
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Human IgE Polyclonal antibody specifically detects Human IgE in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Spezifikation
Human IgE | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Goat | |
Affinity chromatography | |
RUO | |
Human | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1.5 mg/mL | |
PBS with 0.1mM ZnCl2, 1% BSA, 30mM triethanolamine, 5mM MgCl2 and 0.05% sodium azide; pH 7.2 | |
Human IgE | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |