missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG (CH3 domain) Secondary Antibody, Invitrogen™
Mouse Monoclonal Secondary Antibody
Marke: Invitrogen MA190268
Weitere Details : Netto-Gewicht : 1.24740kg
Beschreibung
MA1-90268 detects IgG CH3 domain from human samples.
Human IgG (CH3 domain) Monoclonal antibody specifically detects Human IgG (CH3 domain) in Human samples. It is validated for ELISA, Hemagglutination Assay, Western Blot.Spezifikation
Human IgG (CH3 domain) | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
pFc' from purified protein of Heavy Chain Disease | |
100 μg | |
Secondary | |
4° C, do not freeze | |
IgM |
ELISA, Hemagglutination Assay, Western Blot | |
A57H | |
Unconjugated | |
Mouse | |
purified | |
RUO | |
Human | |
Liquid |