missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgG Fc Highly Cross-Adsorbed Secondary Antibody, FITC, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen A18830
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Human IgG Fc Highly Cross-Adsorbed Polyclonal antibody specifically detects Human IgG Fc Highly Cross-Adsorbed in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.Spezifikation
Human IgG Fc Highly Cross-Adsorbed | |
Polyclonal | |
FITC | |
Goat | |
Affinity chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry, Immunocytochemistry | |
1.5 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.8 | |
Gamma Immunoglobin Fc region | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG |