missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Human IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 755, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen SA510115
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is cross-adsorbed and exhibits minimum reactivity to mouse and rat.
Human IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Human IgG (H+L) Cross-Adsorbed in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Spezifikation
Human IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
DyLight 755 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4 | |
Human IgG-heavy and light chain | |
500 μg | |
Secondary | |
4° C | |
IgG |