missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgG (H+L) Cross-Adsorbed Secondary Antibody, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen A18813
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Human IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Human IgG (H+L) Cross-Adsorbed in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Spezifikation
Human IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Unconjugated | |
Goat | |
Affinity chromatography | |
RUO | |
Human | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
> 4.5 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
Gamma Immunoglobins Heavy and Light chains | |
2 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |