Learn More
F(ab')2-Goat anti-Human IgG, IgM, IgA (H+L) Secondary Antibody, HRP, Invitrogen™
F(ab')2-Goat Polyclonal Secondary Antibody
Marke: Invitrogen A24494
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP). Rehydrate with 0.55 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. Store lyophilized material at 2-8 °C. For long term storage after reconstitution, dilute with 50% glycerol and store at -20 °C as a liquid. Based on Immunoelectrophoresis, no reactivity is observed to: non-immunoglobulin human serum immunoglobulins.
Human IgG, IgM, IgA (H+L) Polyclonal antibody specifically detects Human IgG, IgM, IgA (H+L) in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Spezifikation
Human IgG, IgM, IgA (H+L) | |
Polyclonal | |
HRP | |
Goat | |
Affinity Chromatography | |
RUO | |
Human | |
Lyophilized |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 1% BSA and 0.1% ProClin 150; pH 7.2 | |
Human IgG/A/M | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |